missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Nickel Crucible
Nickel Electro™ Nickel Crucible is crevice free and suitable for temperatures up to 350°C.
Brand: Nickel Electro™ 15
This item is not returnable.
View return policy
Description
- 99.6% nickel purity
- Crevice free
- High thermal and electrical conductivities
- Low gas content
- Low vapor pressure
- Cost effective replacement for platinum
Specifications
Nickel | |
1 mm | |
Autoclavable | |
35 mm |
Each | |
25 mL | |
35 mm | |
Crucible |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction