missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium pieces, 1.0mm (0.04 in.) dia. x 3.18mm (0.125 in.) length, 99% (metals basis), Si nominal 1.5%, Thermo Scientific™

Product Code. 11378388
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

High resistivity material for electronics industry

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11106-97-1
Molecular Formula CrNi
Molecular Weight (g/mol) 110.69
MDL Number MFCD00673110
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
PubChem CID 11320920
IUPAC Name chromium;nickel
SMILES [Cr].[Ni]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Quantity 100 g
Solubility Information Insoluble in water.
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Si nomin.al 1.5%
Odor Odorless
Weight ≈0.21g/piece
Assay (metals basis)
Chemical Name or Material Nickel Chromium pieces, 1.0mm (0.04 in.) dia. x 3.18mm (0.125 in.) length
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.