Learn More
Nickel acetylacetonate, 96%, Thermo Scientific Chemicals
24.32 € - 104.52 €
Chemical Identifiers
CAS | 3264-82-2 |
---|---|
Molecular Formula | C10H14NiO4 |
Molecular Weight (g/mol) | 256.91 |
MDL Number | MFCD00000024 |
InChI Key | BMGNSKKZFQMGDH-FDGPNNRMSA-L |
Synonym | nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii |
PubChem CID | 53384569 |
IUPAC Name | nickel(2+);(E)-4-oxopent-2-en-2-olate |
SMILES | [Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10251123
|
Thermo Scientific Acros
128260050 |
5 g | Glass bottle |
24.32 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10437710
|
Thermo Scientific Acros
128260250 |
25 g | Glass bottle |
64.52 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10371094
|
Thermo Scientific Acros
128261000 |
100 g | Glass bottle |
104.52 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3264-82-2 | |
256.91 | |
BMGNSKKZFQMGDH-FDGPNNRMSA-L | |
53384569 | |
[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
C10H14NiO4 | |
MFCD00000024 | |
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii | |
nickel(2+);(E)-4-oxopent-2-en-2-olate |
Specifications
3264-82-2 | |
100.0 | |
Yellow | |
Authentic | |
C10H14NiO4 | |
05,471; 06,417; 07,250; 08,42; 17,201 | |
nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii | |
BMGNSKKZFQMGDH-FDGPNNRMSA-L | |
nickel(2+);(E)-4-oxopent-2-en-2-olate | |
53384569 | |
96% | |
Nickel acetylacetonate |
95.0 | |
230°C | |
220°C (11.0 mmHg) | |
95% min. (Complexometry) | |
MFCD00000024 | |
15, 6585 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform and benzene,insoluble in ether and ligroin | |
[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
256.91 | |
256.9 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer by inhalation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: I
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 221-875-7
- Nickel acetylacetonate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only