missing translation for 'onlineSavingsMsg'
Learn More

Nickel, 99.9%, (trace metal basis), powder, -325 mesh, Thermo Scientific Chemicals

25.12 € - 193.97 €

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10761661
View Documents
Thermo Scientific Acros
193610250
25 g Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10562791
View Documents
Thermo Scientific Acros
193611000
100 g Plastic bottle
61.20 €
100g
Please to purchase this item. Need a web account? Register with us today!
10003863
View Documents
Thermo Scientific Acros
193615000
500 g Plastic bottle
193.97 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Chemical Identifiers

7440-02-0
58.69
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137 MFCD06798735
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]

Specifications

Brown-Gray
1455.0°C
99.85
8.9
Ni
12,335; 13,197; 17,296
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
58.69
CHEBI:28112
99.9%
Fine Powder
7440-02-0
2730.0°C
100.0
99.9%
MFCD00011137 MFCD06798735
15, 6583
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
935
58.7
Trace Metal Basis
Nickel, 99.9%, -325 mesh
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 231-111-4

missing translation for 'rtecsNumber' : QR5950000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.