missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NH4+ solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
51.20 € - 107.00 €
Specifications
Concentration | 1g/L |
---|---|
Solution Contains | H2O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12992075
|
Fisher Chemical
A 004 100 |
100 mL |
51.20 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12902085
|
Fisher Chemical
A 004 250 |
250 mL |
73.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12912085
|
Fisher Chemical
A 004 500 |
500 mL |
107.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
1g/L | |
H2O |