missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFATC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18222-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located only in the cytoplasm. When translation ends, it interacts with the protein that is a functional homolog of yeast Upf2p to trigger mRNA decapping. Use of multiple polyadenylation sites has been noted for this gene.Specifications
NFATC3 | |
Polyclonal | |
Unconjugated | |
NFATC3 | |
NF AT4, NF ATc3, NFAT4, NFATC3, NFATX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18021, 4775 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97305, Q12968 | |
Nfatc3 | |
NFATC3 Fusion Protein Ag12985 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |