Learn More
NFATC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18222-1-AP-150UL
 View more versions of this product
Product Code. 16848284
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located only in the cytoplasm. When translation ends, it interacts with the protein that is a functional homolog of yeast Upf2p to trigger mRNA decapping. Use of multiple polyadenylation sites has been noted for this gene.
Specifications
Specifications
NFATC3
Polyclonal
Unconjugated
NFATC3
NF AT4, NF ATc3, NFAT4, NFATC3, NFATX
Rabbit
Antigen Affinity Chromatography
RUO
18021, 4775
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97305, Q12968
Nfatc3
NFATC3 Fusion Protein Ag12985
150 μL
Primary
Human, Mouse