Learn More
New Fuchsin, Thermo Scientific Chemicals
38.09 € - 104.52 €
Chemical Identifiers
CAS | 3248-91-7 |
---|---|
Molecular Formula | C22H24ClN3 |
Molecular Weight (g/mol) | 365.91 |
MDL Number | MFCD00012567 |
InChI Key | ORDGVBRMUJCHEO-UHFFFAOYSA-M |
Synonym | Basic Violet 2, C.I. 42520, Magenta III |
PubChem CID | 14454445 |
ChEBI | CHEBI:87671 |
IUPAC Name | 4-[(4-amino-3-methylphenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydrochloride |
SMILES | [Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10082660
|
Thermo Scientific Acros
212140250 |
25 g | Glass bottle |
38.09 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10249463
|
Thermo Scientific Acros
212141000 |
100 g | Glass bottle |
104.52 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3248-91-7 | |
365.91 | |
ORDGVBRMUJCHEO-UHFFFAOYSA-M | |
14454445 | |
4-[(4-amino-3-methylphenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydrochloride |
C22H24ClN3 | |
MFCD00012567 | |
Basic Violet 2, C.I. 42520, Magenta III | |
CHEBI:87671 | |
[Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1 |
Specifications
1500 (at lambda min.) | |
1.1 to 1.4 | |
MFCD00012567 | |
ORDGVBRMUJCHEO-UHFFFAOYSA-M | |
[Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1 | |
365.91 | |
CHEBI:87671 | |
365.89 | |
Crystalline Powder or Crystals |
3248-91-7 | |
C22H24ClN3 | |
Basic Violet 2, C.I. 42520, Magenta III | |
Authentic | |
4-[(4-amino-3-methylphenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydrochloride | |
14454445 | |
550 to 554nm (in 50% ethanol) | |
Green | |
New Fuchsin |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 221-831-7
missing translation for 'rtecsNumber' : CX9825000
missing translation for 'tsca' : TSCA
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H350-May cause cancer.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.