missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neutral Red

Product Code. 15495319
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Nuclear counterstain useful in pH range 6.8 to 8.0

Nuclear counterstain useful in pH range 6.8 - 8.0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-24-2
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) 288.779
MDL Number MFCD00012651
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
Synonym Basic Red 5; C.I. 50040
PubChem CID 11105
ChEBI CHEBI:86370
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl

Specifications

Melting Point 290°C (decomposition)
Beilstein 3918740
Merck Index 14,6488
Formula Weight 288.78
Color Black to Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material Neutral Red

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.