missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeuN Mouse anti-Human, Unconjugated, Clone: 3A4C1, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | NeuN |
---|---|
Clone | 3A4C1 |
Concentration | 1000 μg/mL |
Applications | Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16809085
|
Proteintech
66836-1-IG-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16899075
|
Proteintech
66836-1-IG-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NeuN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeuN. This antibody recognizes human, mouse, rat antigen. The NeuN antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
NeuN | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
146713 | |
Primary |
3A4C1 | |
Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |