missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-Benzyloxycarbonyl-D-lysine, 95%, Thermo Scientific Chemicals
42.70 € - 607.00 €
Chemical Identifiers
CAS | 34404-32-5 |
---|---|
Molecular Formula | C14H20N2O4 |
Molecular Weight (g/mol) | 280.32 |
MDL Number | MFCD00063120 |
InChI Key | CKGCFBNYQJDIGS-STGVRZAANA-N |
Synonym | h-d-lys z-oh, n6-phenylmethoxy carbonyl-r-lysine, r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 2r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, n-e-z-d-lysine, ambotzhaa6320, nepsilon-cbz-d-lysine, d-lys z-oh, nepsilon-carbobenzoxy-d-lysine, n6-benzyloxycarbonyl-d-lysine |
PubChem CID | 1551330 |
IUPAC Name | (2R)-2-amino-6-(phenylmethoxycarbonylamino)hexanoic acid |
SMILES | N[C@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34404-32-5 | |
280.32 | |
CKGCFBNYQJDIGS-STGVRZAANA-N | |
1551330 | |
N[C@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O |
C14H20N2O4 | |
MFCD00063120 | |
h-d-lys z-oh, n6-phenylmethoxy carbonyl-r-lysine, r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 2r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, n-e-z-d-lysine, ambotzhaa6320, nepsilon-cbz-d-lysine, d-lys z-oh, nepsilon-carbobenzoxy-d-lysine, n6-benzyloxycarbonyl-d-lysine | |
(2R)-2-amino-6-(phenylmethoxycarbonylamino)hexanoic acid |
Specifications
34404-32-5 | |
C14H20N2O4 | |
h-d-lys z-oh, n6-phenylmethoxy carbonyl-r-lysine, r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 2r-2-amino-6-benzyloxy carbonyl amino hexanoic acid, n-e-z-d-lysine, ambotzhaa6320, nepsilon-cbz-d-lysine, d-lys z-oh, nepsilon-carbobenzoxy-d-lysine, n6-benzyloxycarbonyl-d-lysine | |
N[C@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O | |
280.32 | |
280.32 | |
Powder |
White | |
MFCD00063120 | |
CKGCFBNYQJDIGS-STGVRZAANA-N | |
(2R)-2-amino-6-(phenylmethoxycarbonylamino)hexanoic acid | |
1551330 | |
95% | |
Nepsilon-Benzyloxycarbonyl-D-lysine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only