Learn More
Ansell Edmont™ Neox™ 09-022 Series Black Neoprene Immersion Gloves
Product Code. 12629721 Shop All Ansell Products
Size 10
Click to view available options
Size 10
Protects against a wide range of oils, acids, caustics, alcohols and many solvents. Special Hi-Lo insulated gauntlet option with quilted cotton liner, for use in both hot and cold temperatures.
  • Permits intermittent handling in hot liquids up to 180°C and handling in cold temperatures down to -25°C.
  • Comfortable anatomical glove shape which follows the contours of the hand at rest.
  • Improves dexterity and reduces hand fatigue.
  • Good resistance to all mechanical hazards.
  • Neoprene Coating.
  • Smooth finish grip design.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Neoprene
Certifications/Compliance Standard CATEGORY III, EN 374, EN 388 : 3121, ANSI, FDA 21 CFR 177.2600
Cuff Material Neoprene
Coating Material Neoprene
Product Type Immersion Gloves
Size 10
Style 09-022
Temperature (Metric) Max 180°C
Glove Cuff Style Gauntlet
Lining Cotton Jersey
Palm Type Fully coated-palm
Quantity 6 Pairs
Resistant To Excellent broad-spectrum chemicals
Series 09-022
Product Title
Ansell Edmont™ Neox™ 09-022 Series Black Neoprene Immersion Gloves >