Learn More
Neopentyl bromide, 98%, Thermo Scientific Chemicals
28.50 € - 181.00 €
Chemical Identifiers
CAS | 630-17-1 |
---|---|
Molecular Formula | C5H11Br |
Molecular Weight (g/mol) | 151.05 |
MDL Number | MFCD00000209 |
InChI Key | CQWYAXCOVZKLHY-UHFFFAOYSA-N |
Synonym | neopentyl bromide, propane, 1-bromo-2,2-dimethyl, 1-bromo-2,2-dimethyl-propane, ccris 4895, neopentylbromde, neopentylbromide, neo-pentyl bromide, ch3 3cch2br, 3-bromo-2,2-dimethylpropyl, neopentyl bromide 5g |
PubChem CID | 12415 |
IUPAC Name | 1-bromo-2,2-dimethylpropane |
SMILES | CC(C)(C)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10470504
|
Thermo Scientific Acros
425720010 |
1 g | Glass bottle |
28.50 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10515134
|
Thermo Scientific Acros
425720050 |
5 g | Glass bottle |
59.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10316774
|
Thermo Scientific Acros
425720250 |
25 g | Glass bottle |
181.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
630-17-1 | |
151.05 | |
CQWYAXCOVZKLHY-UHFFFAOYSA-N | |
12415 | |
CC(C)(C)CBr |
C5H11Br | |
MFCD00000209 | |
neopentyl bromide, propane, 1-bromo-2,2-dimethyl, 1-bromo-2,2-dimethyl-propane, ccris 4895, neopentylbromde, neopentylbromide, neo-pentyl bromide, ch3 3cch2br, 3-bromo-2,2-dimethylpropyl, neopentyl bromide 5g | |
1-bromo-2,2-dimethylpropane |
Specifications
630-17-1 | |
100.0 | |
105°C to 106°C (767.0 mmHg) | |
97.5% min. (GC) | |
(CH3)3CCH2Br | |
1.199 | |
CQWYAXCOVZKLHY-UHFFFAOYSA-N | |
1-bromo-2,2-dimethylpropane | |
12415 | |
98% |
97.5 | |
1.1990g/mL | |
7°C | |
C5H11Br | |
MFCD00000209 | |
neopentyl bromide, propane, 1-bromo-2,2-dimethyl, 1-bromo-2,2-dimethyl-propane, ccris 4895, neopentylbromde, neopentylbromide, neo-pentyl bromide, ch3 3cch2br, 3-bromo-2,2-dimethylpropyl, neopentyl bromide 5g | |
CC(C)(C)CBr | |
151.05 | |
151.04 | |
Neopentyl bromide |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 211-132-5
- Neopentyl bromide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only