Learn More
Neopentyl bromide, 98%, Thermo Scientific Chemicals
65.83 € - 180.90 €
Chemical Identifiers
CAS | 630-17-1 |
---|---|
Molecular Formula | C5H11Br |
Molecular Weight (g/mol) | 151.05 |
MDL Number | MFCD00000209 |
InChI Key | CQWYAXCOVZKLHY-UHFFFAOYSA-N |
Synonym | neopentyl bromide, propane, 1-bromo-2,2-dimethyl, 1-bromo-2,2-dimethyl-propane, ccris 4895, neopentylbromde, neopentylbromide, neo-pentyl bromide, ch3 3cch2br, 3-bromo-2,2-dimethylpropyl, neopentyl bromide 5g |
PubChem CID | 12415 |
IUPAC Name | 1-bromo-2,2-dimethylpropane |
SMILES | CC(C)(C)CBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11352226
|
Thermo Scientific Alfa Aesar
L03579.06 |
5 g |
65.83 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11362226
|
Thermo Scientific Alfa Aesar
L03579.14 |
25 g |
180.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neopentyl bromide is used in synthesis of S-alkylated cysteine derivatives with branched alkyl chains. It can be used in agrochemical, pharmaceutical and dyestuff field .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNeopentyl bromide is used in synthesis of S-alkylated cysteine derivatives with branched alkyl chains. It can be used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Soluble in ethanol, ether, aetone and benzene. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases.
Chemical Identifiers
630-17-1 | |
151.05 | |
CQWYAXCOVZKLHY-UHFFFAOYSA-N | |
12415 | |
CC(C)(C)CBr |
C5H11Br | |
MFCD00000209 | |
neopentyl bromide, propane, 1-bromo-2,2-dimethyl, 1-bromo-2,2-dimethyl-propane, ccris 4895, neopentylbromde, neopentylbromide, neo-pentyl bromide, ch3 3cch2br, 3-bromo-2,2-dimethylpropyl, neopentyl bromide 5g | |
1-bromo-2,2-dimethylpropane |
Specifications
630-17-1 | |
104°C to 106°C | |
C5H11Br | |
MFCD00000209 | |
1730989 | |
Soluble in ethanol,ether,aetone and benzene. Insoluble in water. | |
CC(C)(C)CBr | |
151.05 | |
151.05 | |
Neopentyl bromide |
1.195 | |
6°C (42°F) | |
1.436 | |
UN1993 | |
neopentyl bromide, propane, 1-bromo-2,2-dimethyl, 1-bromo-2,2-dimethyl-propane, ccris 4895, neopentylbromde, neopentylbromide, neo-pentyl bromide, ch3 3cch2br, 3-bromo-2,2-dimethylpropyl, neopentyl bromide 5g | |
CQWYAXCOVZKLHY-UHFFFAOYSA-N | |
1-bromo-2,2-dimethylpropane | |
12415 | |
98% |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 211-132-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.