missing translation for 'onlineSavingsMsg'
Learn More

Neodymium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

100.20 € - 320.60 €

Chemical Identifiers

CAS 334869-71-5
Molecular Formula C6H9NdO6
Molecular Weight (g/mol) 321.37
MDL Number MFCD00013040
InChI Key KETUDCKOKOGBJB-UHFFFAOYSA-K
IUPAC Name neodymium(3+) triacetate
SMILES [Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11445370
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044587.18
50 g
100.20 €
50g
Please to purchase this item. Need a web account? Register with us today!
11455370
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044587.30
250 g
320.60 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Neodymium Acetate, mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

334869-71-5
321.37
KETUDCKOKOGBJB-UHFFFAOYSA-K
[Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
C6H9NdO6
MFCD00013040
neodymium(3+) triacetate

Specifications

334869-71-5
C6H9NdO6
Soluble in water,moderately soluble in strong mineral acids.
[Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
321.37
99.9%
Neodymium (III) acetate hydrate
Crystalline
MFCD00013040
KETUDCKOKOGBJB-UHFFFAOYSA-K
neodymium(3+) triacetate
321.38 (Anhydrous)
(REO)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only