missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16720-1-AP-150UL
This item is not returnable.
View return policy
Description
NDUFB11 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed not to be involved in catalysis. NDUFB11 functions in the transfer of electrons from NADH to the respiratory chain and is located in the inner mitochondrial membrane. The immediate electron acceptor for NDUFB11 is believed to be ubiquinone. Further, NDUFB11 is believed to have NADH dehydrogenase activity, and oxidoreductase activity. Diseases associated with NDUFB11 protein dysfunction include linear skin defects with multiple congenital anomalies and mitochondrial complex I deficiency.Specifications
NDUFB11 | |
Polyclonal | |
Unconjugated | |
NDUFB11 | |
CI ESSS, Complex I ESSS, ESSS, NDUFB11, Neuronal protein 17.3, P17.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104130, 299310, 54539 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O09111, Q9NX14 | |
NDUFB11 | |
NDUFB11 Fusion Protein Ag10159 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |