missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natural Wood Fireproof Stools
149.00 € - 156.00 €
Specifications
Finish | Fireproof |
---|---|
Height (Metric) | 646 mm |
With Backrest | Yes |
Product Type | Stool |
Color | Natural |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11947654
|
mb2
AB4B2D1NE2 |
5 Wheels |
156.00 €
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
11937654
|
mb2
AB4B2D1NE3 |
5 Pads |
149.00 €
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
Specifications
Fireproof | |
Yes | |
Natural |
646 mm | |
Stool | |
Wood |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title