Learn More
Deutsch Neumann™ Vacuum Quality Laboratory Tubing
Provides flexibility, tear strength and resilience for vacuum applications. Deutsch Neumann™ Vacuum Quality Laboratory Tubing is manufactured from red natural rubber.
6.20 €
Specifications
Material Natural Rubber
Overall Length (Metric) 25 m
Color Red
View More Specs
Products 1
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
10509661
View Documents
Deutsch Neumann™
3020717
7 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Durometer 50 Shore A
  • Good elastic memory
  • Excellent choice for vacuum applications
Specifications
Specifications
Natural Rubber
25 m
Red