missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Edmont™ Profil™ Plus 87-850 Series Yellow Natural Rubber Latex Immersion Gloves
Click to view available options
Size:
8
9
Description
- Specially treated to reduce the risk of allergy
- Excellent resistance to mechanical hazards and enhanced chemical resistance
- 100% cotton flocklining - soft, comfortable and absorbent
- Ideal for janitorial use and handling in wet conditions
Specifications
Specifications
Certifications/Compliance | EN420:2003A1:2009, EN374: 2003,EN388: 2003 |
Coating Material | Natural Rubber Latex |
Color | Yellow |
Latex Free | Yes |
Material | Rubber Latex |
Powdered | Powder Free |
Product Type | Gloves |
Size | 9 |
Thickness (Metric) | 0.55 mm |
For Use With (Application) | General handling in wet conditions |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction