Learn More
Thermo Scientific™ National Unassembled Certified™ Vial Kits
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
Brand:  Thermo Scientific™ CERT4000-71LV
 View more versions of this product
Product Code. 11921543
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
  • Fully lot tested for HPLC and GC analysis
  • Certificate of Conformance in every pack
  • Bonded caps to prevent septa push-through
  • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
  • Available with or without ID patch
  • Fixed micro vials or inserts for limited-volume sampling
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Certified Target DP 350µL Clear Glass I-D Fused Insert Vial, 9mm Screw Thread
PTFE, Silica Glass
350 μL
Bonded PTFE, Silicone
9-425
Clear
Yes
PTFE/Silica
Screw Cap
Yes