Learn More
Thermo Scientific™ National Target™ Unassembled Vial, Cap and Septum Convenience Kits
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
33.85 € - 68.66 €
Specifications
Closure Color Light Blue
Volume (Metric) 2 mL
View More Specs
Products 7
Product Code Brand Closure Color Septum Color Marking Spot Price Quantity & Availability  
Product Code Brand Closure Color Septum Color Marking Spot Price Quantity & Availability  
12473678
View Documents
Thermo Scientific™
C4010-88A
Light Blue Red PTFE/White Silicone/Red PTFE Amber No This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12433688
View Documents
Thermo Scientific™
C4010-89AW
Light Blue Red PTFE/White Silicone/Red PTFE Amber Yes This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12443688
View Documents
Thermo Scientific™
C4010-89W
Light Blue Red PTFE/White Silicone/Red PTFE Clear Yes This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12483688
View Documents
Thermo Scientific™
C4010-90W
Light Blue PTFE Unassembled Clear Yes This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12403698
View Documents
Thermo Scientific™
C4010-92
Light Blue PTFE/Silicone, Preslit Clear No This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12413718
View Documents
Thermo Scientific™
C4010-S88
Light Blue PTFE/Silicone Clear No This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12423718
View Documents
Thermo Scientific™
C4010-S92
Light Blue PTFE/Silicone, Pre-slit Clear No This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
  • Two-compartment benchtop storage tray
  • I-D™ vials feature write-on patch with graduations
  • All items remain handy and dust-free
  • Caps with septa are not attached to vials (packed in separate polybag)
Includes:
100 vials and 100 caps with pre-assembled septa
Specifications
Specifications
Light Blue
2 mL