Learn More
Thermo Scientific™ National Target™ Snap-It™ Kits
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
Brand:  Thermo Scientific™ C4011-74
 View more versions of this product
Product Code. 12413798
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
  • 2mL,12 × 32mm vials
  • Choice of vial and preassembled seals with septa
  • Packaged in two compartment benchtop storage trays
  • Include matched quantities of vials and Snap-It caps with prefitted septa
  • All items remain dust-free
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Clear
Yes
PTFE/Silicone, Pre-slit
No
Clear Glass; Septum: PTFE/Silicone, Pre-Slit
PTFE, Silicone
2 mL
PTFE, Silicone