Learn More
Thermo Scientific™ National Microvolume Inserts for 8 x 40mm Shell Vials
Thermo Scientific™ National Microvolume Inserts are available in pulled point or precision point design to suit various application needs.
Brand:  Thermo Scientific™ C4015-96P
 View more versions of this product
Product Code. 12400248
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ National Microvolume Inserts are available in pulled point or precision point design to suit various application needs.
  • Polyspring™ inserts feature a polyethylene spring to center the bottom of the insert and to provide a cushion against needle contact
  • Precision point inserts permit maximum sample extraction
  • Use metal spring with inserts that lack the Polyspring
TRUSTED_SUSTAINABILITY
Specifications
Specifications
300 μL
Precision Point Tip
National
8 x 40 mm Shell Vials