missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Scientific Headspace 20mm Crimp Seals

Thermo Scientific™ National Headspace 20mm Unlined Crimp Seals are compatible with all headspace vials.

Brand:  Thermo Scientific™ C4020-3M

Product Code. 12460288

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ National Headspace 20mm Unlined Crimp Seals are compatible with all headspace vials.

  • Quick, convenient, and minimize contamination
  • 20mm Crimp seals with prefitted septa for headspace vials
  • Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

One-Piece; Tin-plated steel; Unlined
Tinplated Steel
20 mm
Gold
National
Headspace Vials
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers