missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Narrow Neck Reagent Bottle
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Height (Metric):
108 mm
136 mm
170 mm
Description
- HDPE, transparent
- PP tamper-evident screw cap, blue
Specifications
Specifications
Capacity (Metric) | 500 mL |
Height (Metric) | 170 mm |
Length (Metric) | 74 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'