missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals α-Naphtholphthalein, pure, indicator

57.08 € - 590.94 €

Chemical Identifiers

CAS 596-01-0
Molecular Formula C28H18O4
Molecular Weight (g/mol) 418.448
MDL Number MFCD00036202
InChI Key HQHBAGKIEAOSNM-UHFFFAOYSA-N
PubChem CID 68993
IUPAC Name 3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10287153
View Documents Promotion Details
Thermo Scientific Chemicals
190120010
1 g Glass bottle
57.08 €
1g
In Stock
to see stock.
Add to basket
 
10286340
View Documents Promotion Details
Thermo Scientific Chemicals
190120050
5 g Glass bottle
170.85 €
5g
In Stock
to see stock.
Add to basket
 
10093463
View Documents Promotion Details
Thermo Scientific Chemicals
190120250
25 g Glass bottle
590.94 €
25g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

596-01-0
418.448
HQHBAGKIEAOSNM-UHFFFAOYSA-N
3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one
C28H18O4
MFCD00036202
68993
C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O

Specifications

550 min. (at lamda max)
596-01-0
MFCD00036202
15, 6474
Authentic
3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one
68993
418.45
Pure
Beige to Pink or Brown
α-Naphtholphthalein
240.0°C to 250.0°C
C28H18O4
18, I, 384
HQHBAGKIEAOSNM-UHFFFAOYSA-N
C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O
418.448
648 to 654nm (in 0.1N NaOH)
5% max. (110°C, 1 hr.)
from pH 7.3 (pinkish-yellow) to pH 8.7 (greenish--blue)
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 209-875-5

missing translation for 'tsca' : TSCA

Product Identifier
  • a-Naphtholphthalein, indicator

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only