Learn More
Trajan Scientific and Medical™ NanoVolume Syringes
Designed to inject down to 50nL with high precision and accuracy.
Brand:  Trajan Scientific and Medical™ 000353
Product Code. 15298433
  • 204.00 € / Each
Explore more special offers
Description
Description
  • High precision and accuracy for small volume injections
  • Replacement plunger/needle kits
  • Kits are matched and must be replaced as a set
Specifications
Specifications
NanoVolume Syringes
0.155 mm
50 mm
Cone
26 G
0.47 mm
0.5 μL