missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester, 97%, Thermo Scientific Chemicals
123.00 € - 545.00 €
Chemical Identifiers
CAS | 30189-36-7 |
---|---|
Molecular Formula | C20H33N3O8 |
Molecular Weight (g/mol) | 443.50 |
MDL Number | MFCD00057898 |
InChI Key | IQVLXQGNLCPZCL-UGPWUYPHNA-N |
Synonym | boc-lys boc-osu, s-2,5-dioxopyrrolidin-1-yl 2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine hydroxysuccinimide ester, n, a,n, a-di-boc-l-lysine hydroxysuccinimide ester, n,n-di-boc-l-lysine hydroxysuccinimide ester, boc-lys-boc-osu, pubchem14936, boc-lys boc-osu calc. based on dry substance, c/n, 2,5-dioxoazolidinyl 2s-2,6-bis tert-butoxy carbonylamino hexanoate, 2,5-dioxopyrrolidin-1-yl n∼2∼,n∼6∼-bis tert-butoxycarbonyl-l-lysinate |
PubChem CID | 13875534 |
IUPAC Name | (2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate |
SMILES | CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428658
|
Thermo Scientific Alfa Aesar
H62068.06 |
5 g |
123.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15446986
|
Thermo Scientific Alfa Aesar
H62068.14 |
25 g | N/A | ||||||
Description
Nalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
30189-36-7 | |
443.50 | |
IQVLXQGNLCPZCL-UGPWUYPHNA-N | |
13875534 | |
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O |
C20H33N3O8 | |
MFCD00057898 | |
boc-lys boc-osu, s-2,5-dioxopyrrolidin-1-yl 2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine hydroxysuccinimide ester, n, a,n, a-di-boc-l-lysine hydroxysuccinimide ester, n,n-di-boc-l-lysine hydroxysuccinimide ester, boc-lys-boc-osu, pubchem14936, boc-lys boc-osu calc. based on dry substance, c/n, 2,5-dioxoazolidinyl 2s-2,6-bis tert-butoxy carbonylamino hexanoate, 2,5-dioxopyrrolidin-1-yl n∼2∼,n∼6∼-bis tert-butoxycarbonyl-l-lysinate | |
(2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate |
Specifications
184°C | |
White | |
MFCD00057898 | |
boc-lys boc-osu, s-2,5-dioxopyrrolidin-1-yl 2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine hydroxysuccinimide ester, n, a,n, a-di-boc-l-lysine hydroxysuccinimide ester, n,n-di-boc-l-lysine hydroxysuccinimide ester, boc-lys-boc-osu, pubchem14936, boc-lys boc-osu calc. based on dry substance, c/n, 2,5-dioxoazolidinyl 2s-2,6-bis tert-butoxy carbonylamino hexanoate, 2,5-dioxopyrrolidin-1-yl n∼2∼,n∼6∼-bis tert-butoxycarbonyl-l-lysinate | |
IQVLXQGNLCPZCL-UGPWUYPHNA-N | |
(2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate | |
13875534 | |
97% | |
Powder |
30189-36-7 | |
C20H33N3O8 | |
1559007 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O | |
443.50 | |
443.49 | |
Moisture sensitive | |
Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only