missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Septum Closures: Sterile, Bulk Pack, 38-430mm
GREENER_CHOICE

Product Code. 11344465
Click to view available options
Closure Size:
24-415 mm
38-430 mm
No. per Case:
200
240
Fit for Purpose:
Sterile
This item is not returnable. View return policy
This item is not returnable. View return policy

Combine elastomeric sealing of silicone with nonreactive PTFE reagent contact surfaces in these septum closures for media bottles.

Combine the elastomeric sealing of silicone with nonreactive polytetrafluoroethylene reagent contact surfaces in Thermo Scientific™ Nalgene™ Septum Closures, for use with Nalgene™ Sterile PET & PETG media bottles.

Features:
  • Sterile 10-6 SAL
  • No adhesives used to attach septum to high density polyethylene closure
  • Non-cytotoxic and non-pyrogenic
  • Dual septum material for improved gas barrier

Ordering Information:

  • Closures are bulk packed for easy handling.
  • Order Nalgene PET or PETG bottles without closures separately.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Nalgene Closure for Media Bottles
Material HDPE
No. per Case 200
No. per Pack 200
Packaging Bulk
Sterility Sterile
Liner Material Silicone, PTFE
Certifications/Compliance Manufactured in compliance with ISO 13485 quality requirements
Closure Included Yes
Fit for Purpose Sterile
Product Type Septum Closure
For Use With (Equipment) Nalgene Sterile PET and PETG media bottles
Closure Material Polyethylene
Closure Size 24-415 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'