Learn More
Thermo Scientific™ Nalgene™ Autoclavable PPCO Wash Bottles
Reduce the risk of contamination when dispensing sterile water or reagents by using these autoclavable PPCO wash bottles.
64.50 € - 70.35 €
Specifications
Color | Translucent |
---|---|
Includes | Screw/stem and draw tube |
Material | PPCO |
Sterility | Non-sterile |
Closure Included | Yes |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10618101
|
Thermo Scientific™
2405-0500 |
500 mL | 28 mm |
70.35 €
Pack of 6 |
In Stock
Log in to see stock. |
||||
10227830
|
Thermo Scientific™
2405-1000 |
1000 mL | 38-430 mm |
64.50 €
Pack of 4 |
In Stock
Log in to see stock. |
||||
Description
Reduce the risk of contamination when dispensing sterile water or reagents by using autoclavable Thermo Scientific™ Nalgene™ PPCO Wash Bottles. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. These squeeze bottles are flexible, contact-clear, and designed to dispense through the top with a closure and stem molded in one piece for reliable service. Bent stem dispensing tube retains its bend after autoclaving but bend angle can be modified by hand while warm if desired. The tight fit of the draw tube and closure prevents separation when squeezing bottle.
- Autoclavable
- Polypropylene screw/stem and draw tube
- Tip can be cut back to provide increased flow
- Sputter-free, precision stream
- Bottle and Carton are recyclable in many communities
- Manufactured from non-toxic materials
Specifications
Translucent | |
PPCO | |
Yes | |
No | |
Cylindrical | |
Polypropylene | |
Yes |
Screw/stem and draw tube | |
Non-sterile | |
Autoclavable | |
Narrow | |
Wash Bottle | |
Cap |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.