missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
GREENER_CHOICE

Product Code. 10447043
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
20-415 mm
24-415 mm
38-430 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Pack and ship liquid medias, buffers and sera using these heavy-walled, durable PETG square media bottles, designed for use in filling lines.

Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.

Features:
  • Heavy-walled and durable
  • Shatter-resistant
  • Excellent gas barrier properties
  • Sterile

    10-6

    SAL
  • Molded in graduations
  • Closure supplied separately
TRUSTED_SUSTAINABILITY

Specifications

Capacity 2 oz., 60 mL
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Clear
Description Media bottles without closure
Diameter (English) 1.6 in.
Diameter (English) Inner Neck 0.71 in.
Diameter (Metric) Inner Neck 18 mm
For Use With (Application) Filling Lines
Material PETG
No. per Case 200
No. per Pack 40
Sterility Sterile
Closure Included No
Graduated Yes
Mouth Narrow
Shape Square
Fit for Purpose Sterile
Height (Metric) 41 mm
Product Type Media Bottle
Height (English) 1.6 in.
Closure Size 24-415 mm
Diameter (Metric) 41 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.