missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
20-415 mm
24-415 mm
38-430 mm
Fit for Purpose:
Sterile
Description
Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.
Features:
- Heavy-walled and durable
- Shatter-resistant
- Excellent gas barrier properties
- Sterile
10-6
SAL - Molded in graduations
- Closure supplied separately
Specifications
Specifications
Capacity | 4 oz., 125 mL |
Capacity (Metric) | 125 mL |
Capacity (English) | 4 oz. |
Color | Clear |
Description | Media bottles without closure |
Diameter (English) | 2.09 in. |
Diameter (English) Inner Neck | 1.1 in. |
Diameter (Metric) Inner Neck | 28 mm |
For Use With (Application) | Filling Lines |
Material | PETG |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'