Learn More
Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles without Closure: Bulk Pack
Safely store and ship light-sensitive, small-volume, diagnostic samples and valuable reagents with these opaque, amber HDPE bottles.
1226.14 € - 1310.00 €
Specifications
Color | Opaque Amber |
---|---|
Material | HDPE |
No. per Case | 2000 |
No. per Pack | 2000 |
Sterility | Non-sterile |
Product Code | Brand | Capacity (Metric) | Neck Finish | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Neck Finish | Price | Quantity & Availability | ||||
30117593
|
Thermo Scientific™
362004-9125 |
4 mL | 13-415 mm |
1226.14 €
Pack of 2000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10456663
|
Thermo Scientific™
362004-9025 |
8 mL | 20-415 mm |
1242.30 €
Pack of 2000 |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||
10620524
|
Thermo Scientific™
362004-9050 |
15 mL | 20-415 mm |
1310.00 €
Pack of 2000 |
In Stock
Log in to see stock. |
||||
Description
Safely store and ship light-sensitive, diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles, used in test kits and many small-volume applications. These opaque, amber, high-density polyethylene bottles are for use with light-sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Features:- Nonsterile
- Durable, uniform walls resistant to splitting or puncturing
- Leakproof† when used with Nalgene closures (must be ordered separately)
Ordering Information:
Bottles are bulk packed without closures. Packed 2,000 per case. Order colored closures separately (Cat. No. 362150).
Specifications
Opaque Amber | |
2000 | |
Non-sterile | |
Narrow |
HDPE | |
2000 | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition. | |
Sample Container |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.