missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure
GREENER_CHOICE

Perform light-duty, general purpose applications where a heavier bottle is not required with these narrow-mouth PPCO economy bottles with closure.

63.40 € - 107.00 €

Specifications

Color Translucent
Description Narrow-Mouth Economy Bottle
Material PPCO
Sterility Non-sterile
Closure Color Natural
View More Specs

Products 6
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
11399604
GREENER_CHOICE
View Documents
Thermo Scientific™
2087-0001
30 mL 20 mm
67.10 €
Pack of 72
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
 
11309614
GREENER_CHOICE
Vedere i documenti
Thermo Scientific™
2087-0002
60 mL 20 mm
73.10 €
Pack of 72
Estimated Shipment: 15-04-2025
to see stock.
Aggiungi al carrello
 
11319614
GREENER_CHOICE
Vedere i documenti
Thermo Scientific™
2087-0004
125 mL 24 mm
80.90 €
Pack of 72
In Stock
to see stock.
Aggiungi al carrello
 
11329614
GREENER_CHOICE
Vedere i documenti
Thermo Scientific™
2087-0008
250 mL 24 mm
107.00 €
Pack of 72
Estimated Shipment: 04-06-2025
to see stock.
Aggiungi al carrello
 
11339614
GREENER_CHOICE
Vedere i documenti
Thermo Scientific™
2087-0016
500 mL 28 mm
98.55 €
Pack of 48
Estimated Shipment: 17-04-2025
to see stock.
Aggiungi al carrello
 
11349614
Vedere i documenti
Thermo Scientific™
2087-0032
1 L 38-430 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Descrizione

Descrizione

Perform light-duty, general-purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure. These polypropylene copolymer bottles are lightweight versions of the original Nalgene Lab Quality bottle. Made from high quality laboratory grade plastic materials for reliably low leachables and extractables.

Features:
  • Economical and recyclable; designed for single use
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps ensure leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable/Leakproof† when used with Nalgene closures
  • Manufactured from non-toxic materials
Note:
  • Completely disengage threads or remove cap before autoclaving
Includes:
  • Polypropylene closures
.

Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.
Specifica

Specifica

Translucent
PPCO
Natural
Yes
Narrow
Unlined
Screw Cap
Narrow-Mouth Economy Bottle
Non-sterile
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Autoclavable
Round
Polypropylene
No
Videos
SDS
Documenti

Documenti

Product Certifications

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato