Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Freeze samples effectively with impact-resistant these narrow-mouth, polycarbonate bottles with closure.
31.10 € - 90.90 €
Specifications
Color | Clear |
---|---|
Description | Narrow-Mouth Bottle |
Material | Polycarbonate |
Sterility | Non-sterile |
Closure Color | Natural |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10060681
|
Thermo Scientific™
2205-0016 |
0.5 L | 38-430 mm |
47.94 €
Pack of 4 |
In Stock
Log in to see stock. |
||||
10189630
|
Thermo Scientific™
2205-0032 |
1 L | 38-430 mm |
50.83 €
Pack of 4 |
In Stock
Log in to see stock. |
||||
10576922
|
Thermo Scientific™
DS2205-0210 |
2 L | 38-430 mm |
31.10 €
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
||||
10146070
|
Thermo Scientific™
DS2205-0250 |
2.5 L | 38-430 mm |
31.60 €
Each |
In Stock
Log in to see stock. |
||||
10106120
|
Thermo Scientific™
DS2205-0010 |
4 L | 38-430 mm |
57.00 €
Each |
In Stock
Log in to see stock. |
||||
10395841
|
Thermo Scientific™
DS2205-0020 |
8 L | 53B |
90.90 €
Each |
In Stock
Log in to see stock. |
||||
Description
Freeze samples effectively with impact-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure, which are window clear. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features:- 16 oz. and 0.25 gal. sizes are good for smaller volume applications where contents need to be seen
- 0.5 and 0.66 gal. sizes can be used as culture vessels for batch, suspension or nonattached roller culturing
- 1 and 2 gal. sizes have built-in shoulder loops for attaching an identification tag
- Excellent impact resistance makes these bottles good for freezer use to -135°C (-211°F)
- Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
- Autoclavable/Transparent/Leakproof† when used with Nalgene closures
Includes:
Leakproof† polypropylene screw caps
Note:
Before autoclaving, completely disengage threads or remove cap. Autoclave empty. Repeated autoclaving will shorten the bottle life.
.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged.
Specifications
Clear | |
Polycarbonate | |
Natural | |
Yes | |
Narrow | |
Narrow Mouth Bottle | |
Polypropylene |
Narrow-Mouth Bottle | |
Non-sterile | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1580 for food and beverage use, USP Class VI. | |
Autoclavable | |
Round | |
Unlined | |
No |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.