missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE, Round Carboy

For Use With: Store and transport reagents; Closure Material: Polypropylene; Color: White

111.00 € - 360.00 €

Specifications

Autoclavable Not Autoclavable
Closure Material Polypropylene
Closure Size 83B
Color White
Description Round LDPE Carboy with Handle
View More Specs

Products 5
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10670714
View accessory products View Documents
Thermo Scientific™
2210-0020
10 L
360.00 €
Pack of 6
In Stock
to see stock.
Edge
Add to basket
 
15971481
View Documents
Thermo Scientific™
2210-0040
15 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10250831
View accessory products View Documents
Thermo Scientific™
2210-0050
20 L
154.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
10792701
View accessory products View Documents
Thermo Scientific™
2210-0065
25 L
111.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
10119170
View accessory products View Documents
Thermo Scientific™
2210-0130
50 L
317.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.

  • Molded-in graduations of 5 L and 1 gal. increments
  • Leakproof†

Includes:White polypropylene screw cap and 83B closure

Specifications

Specifications

Not Autoclavable
83B
Round LDPE Carboy with Handle
Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
LDPE
Round
Polypropylene
White
64 mm
Handles
Carboy with Handle
Sterile
Videos
SDS
Documents

Documents

Product Certifications

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.