missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE, Single-Use Sterile Carboy

For Use With: Ideal for storing sterile fluids and pharmaceutical/biotech products.; Closure Material: Polypropylene; Color: White; Sterility: Sterile

272.00 € - 1005.00 €

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Food-grade, USP Class VI, non-cytotoxic
Closure Material Polypropylene
Closure Size 83B
Color White
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10758851
View Documents Promotion Details
Thermo Scientific™
342289-0050
20 L
1005.00 €
Pack of 6
In Stock
to see stock.
Edge
Add to basket
 
15430655
View Documents Promotion Details
Thermo Scientific™
342289-0075
33 L
272.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Eliminate cleaning, prep and set-up costs with ready-to-use Thermo Scientific™ Nalgene™ Single-Use HDPE Carboys. Supplied sterile for one-time use, the high-density polyethylene vessels are ideal for storing sterile fluids and pharmaceutical/biotech products.

Features

  • HDPE carboy with polypropylene closure
  • Sterile
  • Leakproof†

Ordering Information

Attachable stainless-steel handle available (Cat. No. 2229-0001)

Includes

Polypropylene (PP) closure

Specifications

Specifications

Not Autoclavable
Polypropylene
White
Single-use
HDPE
Sterile
Food-grade, USP Class VI, non-cytotoxic
83B
64 mm
Storage, Transportation ​
Carboy
Videos
SDS
Documents

Documents

Product Certifications

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.