Learn More
Thermo Scientific™ Nalgene™ HDPE, Rectangular Carboy
For Use With: Convenient for dry materials such as solids and powders; Closure Material: Polypropylene; Color: White
87.15 € - 129.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
Closure Material | Polypropylene |
Closure Size | 100-415 mm |
Color | White |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10706801
|
Thermo Scientific™
2211-0020 |
9 L |
87.15 €
Each |
In Stock
Log in to see stock. |
|||||
10180341
|
Thermo Scientific™
2211-0050 |
20 L |
129.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Easily fill, transfer and clean using the wide-mouth opening of Thermo Scientific™ Nalgene™ HDPE Rectangular Carboys without Spigots. Featuring a sturdy, space-saving design, these high density polyethylene carboys have convenient molded-in graduations to deliver in liters and gallons.
- Graduated to contain in 1 gal. and 5 L increments for ease of us
- Convenient for dry materials such as solids and powders
- Leakproof†
Includes:
- Polypropylene 100 mm screw closures
Specifications
Not Autoclavable | |
100-415 mm | |
Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation | |
1 Ea. | |
Sterile |
Polypropylene | |
White | |
HDPE | |
Rectangular Carboy |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for two minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.