missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottle

Product Code. 16450631
Click to view available options
Capacity (Metric):
5 mL
20 mL
1 L
2 L
5 L
10 L
Shape:
Round
Square
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottles are certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination.

Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations
  • Durable, translucent PC construction ensures the integrity of your sample
  • Designed specifically for pharmaceutical and biological manufacturers
  • Closures feature a silicone liner for leakproof assurance
  • Extensive validation documentation support
  • Excellent for freezing applications
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 L
Capacity (English) 169 oz.
Color Clear
Diameter (Metric) Inner Neck 48 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Height (Metric) With Closure 299 mm
Material Polycarbonate
No. per Case 6/Cs.
Sterility Sterile
Temperature Range (Metric) -100°C to 100°C
Weight (English) 8.6 lb.
Closure Color White
Low Particulate <1/3 USP <788> (Ultralow particulate)
Certifications/Compliance < 1/3 particulates allowed according to USP <778>
Closure Included Yes
Autoclavable Not Autoclavable
Mouth Narrow
Shape Square
Height (Metric) 299 mm
Product Type Plastic Media Bottle
Width (English) 14.3 in.
Closure Liner Silicone
Closure Material Polypropylene
Closure Size 48 mm
Diameter (Metric) 166 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'