missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottle
Click to view available options
Capacity (Metric):
5 mL
20 mL
1 L
2 L
5 L
10 L
Shape:
Round
Square
Description
Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.
- Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations
- Durable, translucent PC construction ensures the integrity of your sample
- Designed specifically for pharmaceutical and biological manufacturers
- Closures feature a silicone liner for leakproof assurance
- Extensive validation documentation support
- Excellent for freezing applications
Specifications
Specifications
Capacity (Metric) | 1 L |
Capacity (English) | 33.8 oz. |
Color | Clear |
Diameter (English) Inner Neck | 1.89 in. |
Diameter (Metric) Inner Neck | 48 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product |
Material | Polycarbonate |
No. per Case | 35/Cs. |
No. per Pack | 5/Pk. |
Sterility | Sterile |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'