missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)dimethylamine, 95%, Thermo Scientific Chemicals

51.96 € - 182.91 €

Chemical Identifiers

CAS 2083-91-2
Molecular Formula C5H15NSi
Molecular Weight (g/mol) 117.27
MDL Number MFCD00008297
InChI Key KAHVZNKZQFSBFW-UHFFFAOYSA-N
Synonym n,n-dimethyltrimethylsilylamine, n,n-dimethylaminotrimethylsilane, silanamine, pentamethyl, dimethylamino trimethylsilane, pentamethylsilylamine, n-trimethylsilyl dimethylamine, tmsdma, dimethylaminotrimethylsilane, silanamine, n,n,1,1,1-pentamethyl, n,n-dimethyltrimethylsilamine
PubChem CID 74965
IUPAC Name N-methyl-N-trimethylsilylmethanamine
SMILES CN(C)[Si](C)(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11407877
View Documents
Thermo Scientific Alfa Aesar
A16550.09
10 g
51.96 €
10g
Please to purchase this item. Need a web account? Register with us today!
11417877
View Documents
Thermo Scientific Alfa Aesar
A16550.18
50 g
182.91 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N,N-Dimethyltrimethylsilylamine was used in the synthesis of phosphoramidite. It was also employed as a reagent in the preparation of iminium salts and amides as well as for the silylation of polymers. A combination of N-(trimethylsilyl)dimethylamine and MeI was also effective to give p-methylbenzoic acid with a 85% yield based on 90% conversion from the corresponding Me ester. DETA has been treated with N-(trimethylsilyl)-dimethylamine to yield mono-, di- and tri- silylated derivatives

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyltrimethylsilylamine was used in the synthesis of phosphoramidite. It was also employed as a reagent in the preparation of iminium salts and amides as well as for the silylation of polymers. A combination of N-(trimethylsilyl)dimethylamine and MeI was also effective to give p-methylbenzoic acid with a 85% yield based on 90% conversion from the corresponding Me ester. DETA has been treated with N-(trimethylsilyl)-dimethylamine to yield mono-, di- and tri- silylated derivatives

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications

Chemical Identifiers

2083-91-2
117.27
KAHVZNKZQFSBFW-UHFFFAOYSA-N
74965
CN(C)[Si](C)(C)C
C5H15NSi
MFCD00008297
n,n-dimethyltrimethylsilylamine, n,n-dimethylaminotrimethylsilane, silanamine, pentamethyl, dimethylamino trimethylsilane, pentamethylsilylamine, n-trimethylsilyl dimethylamine, tmsdma, dimethylaminotrimethylsilane, silanamine, n,n,1,1,1-pentamethyl, n,n-dimethyltrimethylsilamine
N-methyl-N-trimethylsilylmethanamine

Specifications

86°C to 87°C
C5H15NSi
MFCD00008297
1731861
n,n-dimethyltrimethylsilylamine, n,n-dimethylaminotrimethylsilane, silanamine, pentamethyl, dimethylamino trimethylsilane, pentamethylsilylamine, n-trimethylsilyl dimethylamine, tmsdma, dimethylaminotrimethylsilane, silanamine, n,n,1,1,1-pentamethyl, n,n-dimethyltrimethylsilamine
KAHVZNKZQFSBFW-UHFFFAOYSA-N
N-methyl-N-trimethylsilylmethanamine
74965
95%
1.397
0.732
2083-91-2
(CH3)3SiN(CH3)2
UN2733
−19°C (−3°F)
Hydrolyzes in water.
CN(C)[Si](C)(C)C
117.27
117.27
Amine-like
Moisture sensitive
N-(Trimethylsilyl)dimethylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 218-222-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.