Learn More
n-Tetracosane, 99%, Thermo Scientific Chemicals
24.10 € - 126.00 €
Chemical Identifiers
CAS | 646-31-1 |
---|---|
Molecular Formula | C24H50 |
Molecular Weight (g/mol) | 338.664 |
MDL Number | MFCD00009352 |
InChI Key | POOSGDOYLQNASK-UHFFFAOYSA-N |
Synonym | n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b |
PubChem CID | 12592 |
ChEBI | CHEBI:32936 |
IUPAC Name | tetracosane |
SMILES | CCCCCCCCCCCCCCCCCCCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348978
|
Thermo Scientific Alfa Aesar
042462.06 |
5 g |
24.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328978
|
Thermo Scientific Alfa Aesar
042462.14 |
25 g |
43.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338978
|
Thermo Scientific Alfa Aesar
042462.22 |
100 g |
126.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Tetracosane is a suitable solvent used in the synthesis of ZnS nanoparticle. It is also employed as the wax component to study the reduction in pour point of hydrocarbon solvents containing wax crystals on addition of polymer additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
646-31-1 | |
338.664 | |
POOSGDOYLQNASK-UHFFFAOYSA-N | |
12592 | |
tetracosane |
C24H50 | |
MFCD00009352 | |
n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b | |
CHEBI:32936 | |
CCCCCCCCCCCCCCCCCCCCCCCC |
Specifications
646-31-1 | |
0.799 | |
>110°C (230°F) | |
MFCD00009352 | |
Insoluble in water. | |
CCCCCCCCCCCCCCCCCCCCCCCC | |
338.664 | |
CHEBI:32936 | |
99% | |
n-Tetracosane |
49°C to 52°C | |
391°C | |
C24H50 | |
n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b | |
POOSGDOYLQNASK-UHFFFAOYSA-N | |
tetracosane | |
12592 | |
338.66 | |
Crystalline |
Safety and Handling
missing translation for 'einecsNumber' : 211-474-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only