Learn More
N-tert-Butylmethylamine, 97%, Thermo Scientific Chemicals
32.40 € - 85.90 €
Chemical Identifiers
CAS | 14610-37-8 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.166 |
MDL Number | MFCD00042853 |
InChI Key | ZQGJEUVBUVKZKS-UHFFFAOYSA-N |
Synonym | n-methyl-tert-butylamine, n-tert-butylmethylamine, 2-propanamine, n,2-dimethyl, tert-butylmethylamine, tert-butyl methyl amine, n-methyl-tert.-butylamine, 2-propanamine,n,2-dimethyl, t-butylmethylamine, methyl-t-butylamine, n-t-butylmethylamine |
PubChem CID | 84552 |
IUPAC Name | N,2-dimethylpropan-2-amine |
SMILES | CC(C)(C)NC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11312259
|
Thermo Scientific Alfa Aesar
H27242.03 |
1 g |
32.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11322259
|
Thermo Scientific Alfa Aesar
H27242.06 |
5 g |
85.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14610-37-8 | |
87.166 | |
ZQGJEUVBUVKZKS-UHFFFAOYSA-N | |
84552 | |
CC(C)(C)NC |
C5H13N | |
MFCD00042853 | |
n-methyl-tert-butylamine, n-tert-butylmethylamine, 2-propanamine, n,2-dimethyl, tert-butylmethylamine, tert-butyl methyl amine, n-methyl-tert.-butylamine, 2-propanamine,n,2-dimethyl, t-butylmethylamine, methyl-t-butylamine, n-t-butylmethylamine | |
N,2-dimethylpropan-2-amine |
Specifications
14610-37-8 | |
−18°C (0°F) | |
C5H13N | |
MFCD00042853 | |
n-methyl-tert-butylamine, n-tert-butylmethylamine, 2-propanamine, n,2-dimethyl, tert-butylmethylamine, tert-butyl methyl amine, n-methyl-tert.-butylamine, 2-propanamine,n,2-dimethyl, t-butylmethylamine, methyl-t-butylamine, n-t-butylmethylamine | |
CC(C)(C)NC | |
87.166 | |
87.16 | |
N-tert-Butylmethylamine |
67°C to 69°C | |
Amine-like | |
(CH3)3CNHCH3 | |
UN2733 | |
ZQGJEUVBUVKZKS-UHFFFAOYSA-N | |
N,2-dimethylpropan-2-amine | |
84552 | |
97% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P210-P280-P305+P351+P338-P309-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
RUO – Research Use Only