missing translation for 'onlineSavingsMsg'
Learn More

n-Propyltriethoxysilane, 97%, Thermo Scientific Chemicals

36.70 € - 406.02 €

Chemical Identifiers

CAS 2550-02-9
MDL Number MFCD00026756
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11450077
View Documents
Thermo Scientific Alfa Aesar
B20997.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11460077
View Documents
Thermo Scientific Alfa Aesar
B20997.22
100 g
107.54 €
100g
Please to purchase this item. Need a web account? Register with us today!
11470077
View Documents
Thermo Scientific Alfa Aesar
B20997.36
500 g
406.02 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Store away from air and oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Specifications

Chemical Identifiers

2550-02-9
MFCD00026756

Specifications

179°C to 180°C
C9H22O3Si
UN1993
57°C (134°F)
NBXZNTLFQLUFES-UHFFFAOYSA-N
triethoxy(propyl)silane
75691
97%
2550-02-9
MFCD00026756
1748158
propyltriethoxysilane, triethoxy propyl silane, n-propyltriethoxysilane, silane, triethoxypropyl, unii-f17otx9fg5, f17otx9fg5, dynasylan pteo, triethoxypropyl-silan, triethoxypropyl-silane, triethoxy-n-propylsilane
CCC[Si](OCC)(OCC)OCC
206.36
206.36
n-Propyltriethoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only