missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyldiethanolamine, 95%, Thermo Scientific™

18.20 € - 85.88 €

Chemical Identifiers

CAS 120-07-0, 62-53-3
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD00002845
InChI Key OJPDDQSCZGTACX-UHFFFAOYSA-N
Synonym n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
PubChem CID 8416
IUPAC Name 2-[N-(2-hydroxyethyl)anilino]ethanol
SMILES OCCN(CCO)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10254750
View Documents
Thermo Scientific Acros
130495000
500 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10675372
View Documents
Thermo Scientific Acros
130490010
1 kg Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11325066
View Documents
Thermo Scientific Acros
130490025
2.5 kg Glass bottle
85.88 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

120-07-0, 62-53-3
181.24
OJPDDQSCZGTACX-UHFFFAOYSA-N
8416
OCCN(CCO)C1=CC=CC=C1
C10H15NO2
MFCD00002845
n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
2-[N-(2-hydroxyethyl)anilino]ethanol

Specifications

120-07-0, 62-53-3
100.0
White to Brown
200°C
94% min. (GC)
(HOCH2CH2)2NC6H5
n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
OJPDDQSCZGTACX-UHFFFAOYSA-N
2-[N-(2-hydroxyethyl)anilino]ethanol
8416
181.23
Crystalline Mass, Crystals or Chunks
94.0
55°C to 59°C
270°C
Authentic
C10H15NO2
MFCD00002845
Solubility in water: 45g/L (20°C). Other solubilities: freely soluble in ethanol,acetone,ethyl acetate,and ether (20°C)
OCCN(CCO)C1=CC=CC=C1
181.24
1.27 mPa.s (200°C)
95%
N-Phenyldiethanolamine, 90-95%
Videos
Seguridad y manipulación

Seguridad y manipulación

GHS H Statement:
Harmful to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-368-5

missing translation for 'rtecsNumber' : KM2100000

missing translation for 'tsca' : TSCA

Recommended Storage : This product tends to darken during storage

Nombre del producto
  • N-Phenyldiethanolamine

Palabra de advertencia
  • Danger

Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Indicaciones de peligro
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.

Consejos de prudencia
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates

RUO – Research Use Only