Learn More
N-Phenylcarbazole, 97%, Thermo Scientific Chemicals
25.42 € - 331.76 €
Chemical Identifiers
CAS | 1150-62-5 |
---|---|
Molecular Formula | C18H13N |
Molecular Weight (g/mol) | 243.31 |
MDL Number | MFCD00004965 |
InChI Key | VIJYEGDOKCKUOL-UHFFFAOYSA-N |
PubChem CID | 70851 |
IUPAC Name | 9-phenylcarbazole |
SMILES | C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1150-62-5 | |
243.31 | |
VIJYEGDOKCKUOL-UHFFFAOYSA-N | |
9-phenylcarbazole |
C18H13N | |
MFCD00004965 | |
70851 | |
C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Specifications
1150-62-5 | |
96% min. (GC) | |
C18H13N | |
20,I,164 | |
VIJYEGDOKCKUOL-UHFFFAOYSA-N | |
9-phenylcarbazole | |
70851 | |
97% |
94.0°C to 97.0°C | |
Glass Bottle | |
MFCD00004965 | |
n-phenylcarbazole, 9-phenyl-9h-carbazole, 9h-carbazole, 9-phenyl, carbazole, 9-phenyl, carbazol, 9-phenyl, 9-phenyl carbazole, pubchem23849, 9h-carbazole,9-phenyl, 9-phenylcarbazole, acmc-2099pd | |
C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2 | |
243.31 | |
243.31 | |
N-Phenylcarbazole |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 214-564-2
- 9-Phenylcarbazole, 97%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only