missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Click to view available options
Quantity:
50 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
n-Pentylzinc bromide used as a intermediate for pharmaceutical.
Solubility
Reacts with water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry an
n-Pentylzinc bromide used as a intermediate for pharmaceutical.
Solubility
Reacts with water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry an
Specifications
Specifications
CAS | 308796-10-3 |
Density | 0.965 |
Molecular Formula | C5H11BrZn |
Linear Formula | CH3(CH2)4ZnBr |
MDL Number | MFCD00671998 |
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air sensitive |
Synonym | pentylzinc bromide solution, bromozinc 1+ ; pentane, bromozincylium; pentan-1-ide |
Solubility Information | Reacts with water. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'