missing translation for 'onlineSavingsMsg'
Learn More

N-Pentane, Certified AR for Analysis, Fisher Chemical™

C5H12, CAS Number-109-66-0, 2.5L, 36 deg.C, CHEBI:37830, Undesignated, 72.15g/mol, OFBQJSOFQDEBGM-UHFFFAOYSA-N, pentane, -130 deg.C, 9498, 72.151, Amber glass bottle, Liquid, 8003, CCCCC, 573mbar at 20 deg.C, 0.25 mPaS at 20 deg.C

83.75 € - 257.00 €

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10366000
SDS View Documents
Fisher Chemical
P/1021/15
1 L Amber glass bottle
83.75 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10558250
SDS View Documents
Fisher Chemical
P/1021/17
2.5 L Amber glass bottle
257.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11458433
SDS View Documents
Fisher Chemical
P/1021/PB17
2.5 L Plastic Coated Glass bottle
248.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C5H12, CAS Number-109-66-0, 2.5L, 36 deg.C, CHEBI:37830, Undesignated, 72.15g/mol, OFBQJSOFQDEBGM-UHFFFAOYSA-N, pentane, -130 deg.C, 9498, 72.151, Amber glass bottle, Liquid, 8003, CCCCC, 573mbar at 20 deg.C, 0.25 mPaS at 20 deg.C

Specifications

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
CHEBI:37830
CCCCC
C5H12
MFCD00009498
8003
pentane

Specifications

109-66-0
0.25 mPaS at 20°C
C5H12
MFCD00009498
36°C
CCCCC
72.15
CHEBI:37830
573mbar at 20°C
2.5
Liquid
-130°C
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Pentane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P273-Avoid release to the environment.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Pentane, Certified AR for Analysis, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.