Learn More
N,N,N',N'-Tetraethylethylenediamine, 99+%, Thermo Scientific Chemicals
48.70 € - 1278.00 €
Chemical Identifiers
CAS | 150-77-6 |
---|---|
Molecular Formula | C10H26N2 |
Molecular Weight (g/mol) | 174.33 |
MDL Number | MFCD00009055 |
InChI Key | DIHKMUNUGQVFES-UHFFFAOYSA-P |
Synonym | n,n,n',n'-tetraethylethylenediamine, teeda, 1,2-ethanediamine, n,n,n',n'-tetraethyl, tetraethylethylenediamine, ethylenediamine, n,n,n',n'-tetraethyl, 1,2-ethanediamine,n1,n1,n2,n2-tetraethyl, n,n,n',n'-tetraethylenediamine, 1,2-ethanediamine, n1,n1,n2,n2-tetraethyl, 2-diethylamino ethyl diethylamine, acmc-20ak9v |
PubChem CID | 67423 |
SMILES | CC[NH+](CC)CC[NH+](CC)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10307860
|
Thermo Scientific Acros
155590100 |
10 mL |
48.70 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10133590
|
Thermo Scientific Acros
155591000 |
100 mL |
256.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11925191
|
Thermo Scientific Acros
155595000 |
500 mL |
1278.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
150-77-6 | |
174.33 | |
DIHKMUNUGQVFES-UHFFFAOYSA-P | |
67423 |
C10H26N2 | |
MFCD00009055 | |
n,n,n',n'-tetraethylethylenediamine, teeda, 1,2-ethanediamine, n,n,n',n'-tetraethyl, tetraethylethylenediamine, ethylenediamine, n,n,n',n'-tetraethyl, 1,2-ethanediamine,n1,n1,n2,n2-tetraethyl, n,n,n',n'-tetraethylenediamine, 1,2-ethanediamine, n1,n1,n2,n2-tetraethyl, 2-diethylamino ethyl diethylamine, acmc-20ak9v | |
CC[NH+](CC)CC[NH+](CC)CC |
Specifications
150-77-6 | |
0.8080g/mL | |
58°C | |
99% min. (GC) | |
C10H26N2 | |
1.4333 to 1.4353 | |
04,251 | |
n,n,n',n'-tetraethylethylenediamine, teeda, 1,2-ethanediamine, n,n,n',n'-tetraethyl, tetraethylethylenediamine, ethylenediamine, n,n,n',n'-tetraethyl, 1,2-ethanediamine,n1,n1,n2,n2-tetraethyl, n,n,n',n'-tetraethylenediamine, 1,2-ethanediamine, n1,n1,n2,n2-tetraethyl, 2-diethylamino ethyl diethylamine, acmc-20ak9v | |
DIHKMUNUGQVFES-UHFFFAOYSA-P | |
N,N,N',N'-tetraethylethane-1,2-diamine | |
67423 | |
99+% | |
N,N,N',N'-Tetraethylethylenediamine |
Yellow | |
189.0°C to 192.0°C | |
Authentic | |
Glass bottle | |
(C2H5)2NCH2CH2N(C2H5)2 | |
MFCD00009055 | |
0.808 | |
Solubility in water: soluble. | |
CC[NH+](CC)CC[NH+](CC)CC | |
174.33 | |
172.31 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-770-3
RUO – Research Use Only