missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, ≥99.8%, ACS Reagent, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
Melting Point | -61°C |
Density | 0.944g/mL |
Boiling Point | 153°C |
Quantity | 1 L |
Explosion Limit | 0.152 |
Flash Point | 58°C (136.4°F) |
Ignition Point | 833°F |
Packaging | Glass Bottle |
Linear Formula | HCON(CH3)2 |
Vapor Pressure | 2.7mmHg (20°C) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'